Tisk…

4-Methoxy-2-methyl-1H-indole 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Methoxy-2-methy-1H-indole

OR951069-1G OR951069-250MG
APOSOR951069-1GEA 9490 CZK
APOSOR951069-1G APOSOR951069-250MG
4-Methoxy-2-methyl-1H-indole 95%
4-Methoxy-2-methyl-1H-indole
Vzorec: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040252
CAS číslo: 17897-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR