Tisk…

6-Nitro-1,2,3,4-tetrahydroquinoline 96%

Dodavatel: Apollo Scientific

OR951068-1G
APOSOR951068-1GEA 2730 CZK
APOSOR951068-1G
6-Nitro-1,2,3,4-tetrahydroquinoline 96%
6-Nitro-1,2,3,4-tetrahydroquinoline
Vzorec: C₉H₁₀N₂O₂
MW: 178,188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00541472
CAS číslo: 14026-45-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR