Tisk…
1-Isobutylpiperidin-4-amine hydrochloride 98%
Dodavatel: Apollo Scientific
OR951050-1G OR951050-5G
APOSOR951050-1GEA 9330 CZK
APOSOR951050-1G APOSOR951050-5G
1-Isobutylpiperidin-4-amine hydrochloride 98%
1-Isobutylpiperidin-4-amine hydrochloride
Vzorec: C₉H₂₀N₂·ClH
MW: 192,729 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054662
CAS číslo: 1177306-12-5