Tisk…

4'-Diethylaminophenyl acetylene 95%

Dodavatel: Apollo Scientific

Synonyms: N,N-Diethyl-4-ethynylaniline

OR951025-1G OR951025-250MG
APOSOR951025-1GEA 9890 CZK
APOSOR951025-1G APOSOR951025-250MG
4'-Diethylaminophenyl acetylene 95%
4'-Diethylaminophenyl acetylene
Vzorec: C₁₂H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD00168817
CAS číslo: 41876-70-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR