Tisk…

1,3-Bis(methylsulphonyl)benzene

Dodavatel: Apollo Scientific
Varování

OR9510-1G OR9510-5G
APOSOR9510-1GEA 1820 CZK
APOSOR9510-1G APOSOR9510-5G
1,3-Bis(methylsulphonyl)benzene
1,3-Bis(methylsulphonyl)benzene
Vzorec: C₈H₁₀O₄S₂
MW: 234,293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08543442
CAS číslo: 22821-85-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR