5-P-Tolylpyridine-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR950985-1G
OR950985-250MG
APOSOR950985-1GEA
21490
CZK
APOSOR950985-1G
APOSOR950985-250MG
5-P-Tolylpyridine-3-carbaldehyde 95%
5-P-Tolylpyridine-3-carbaldehyde
Vzorec:
C₁₃H₁₁NO MW: 197,233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06410229 CAS číslo: 229008-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...