Tisk…

(1-Pyridin-2-ylpropyl)amine 95%

Dodavatel: Apollo Scientific

OR950974-1G OR950974-5G
APOSOR950974-1GEA 2150 CZK
APOSOR950974-1G APOSOR950974-5G
(1-Pyridin-2-ylpropyl)amine 95%
(1-Pyridin-2-ylpropyl)amine
Vzorec: C₈H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD09802324
CAS číslo: 100155-73-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR