Benzyl 2-cyclopropylacetate 95%
Dodavatel: Apollo Scientific
OR950902-1G
OR950902-250MG
APOSOR950902-1GEA
17990
CZK
APOSOR950902-1G
APOSOR950902-250MG
Benzyl 2-cyclopropylacetate 95%
Benzyl 2-cyclopropylacetate
Vzorec:
C₁₂H₁₄O₂ MW: 190,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11976040 CAS číslo: 59698-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...