[1,2,4]triazolo[1,5-a]pyrazin-2-amine 97%
Dodavatel: Apollo Scientific
OR950865-1G
OR950865-250MG
APOSOR950865-1GEA
1420
CZK
APOSOR950865-1G
APOSOR950865-250MG
[1,2,4]triazolo[1,5-a]pyrazin-2-amine 97%
[1,2,4]triazolo[1,5-a]pyrazin-2-amine
Vzorec:
C₅H₅N₅ Storage Temperature: Ambient |
MDL Number:
MFCD11847742 CAS číslo: 88002-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...