Tisk…

Isopropyl cyclopropanesulphonate 95%

Dodavatel: Apollo Scientific

OR950828-1G OR950828-5G
APOSOR950828-1GEA 1020 CZK
APOSOR950828-1G APOSOR950828-5G
Isopropyl cyclopropanesulphonate 95%
Isopropyl cyclopropanesulphonate
Vzorec: C₆H₁₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD21606956
CAS číslo: 146475-51-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR