Tisk…

(S)-b-Amino-4-bromo-benzeneethanol 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-2-Amino-2-(4-bromophenyl)ethanol

OR950823-1G OR950823-250MG
APOSOR950823-1GEA 3910 CZK
APOSOR950823-1G APOSOR950823-250MG
(S)-b-Amino-4-bromo-benzeneethanol 97%
(S)-b-Amino-4-bromo-benzeneethanol
Vzorec: C₈H₁₀BrNO
MW: 216,075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09256372
CAS číslo: 354153-65-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR