Ethyl 2-formylbenzoate 95%
Dodavatel: Apollo Scientific
OR950665-1G
OR950665-250MG
APOSOR950665-1GEA
19440
CZK
APOSOR950665-1G
APOSOR950665-250MG
Ethyl 2-formylbenzoate 95%
Ethyl 2-formylbenzoate
Vzorec:
C₁₀H₁₀O₃ MW: 178.185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09743805 CAS číslo: 34046-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...