2-Amino-6-chlorobenzaldehyde 95%
Dodavatel: Apollo Scientific
OR950569-1G
OR950569-250MG
APOSOR950569-1GEA
3640
CZK
APOSOR950569-1G
APOSOR950569-250MG
2-Amino-6-chlorobenzaldehyde 95%
2-Amino-6-chlorobenzaldehyde
Vzorec:
C₇H₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD10696880 CAS číslo: 35490-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...