Tisk…
1,5-Dimethyl-1H-pyrazol-4-amine dihydrochloride 95%
Dodavatel: Apollo Scientific
OR950407-1G OR950407-250MG
APOSOR950407-1GEA 11810 CZK
APOSOR950407-1G APOSOR950407-250MG
1,5-Dimethyl-1H-pyrazol-4-amine dihydrochloride 95%
1,5-Dimethyl-1H-pyrazol-4-amine dihydrochloride
Vzorec: C₅H₉N₃·2ClH
MW: 184.067 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969122
CAS číslo: 1189950-55-7