Tisk…

1-(2-Bromobenzyl)-1H-imidazole ≥98%

Dodavatel: Apollo Scientific

OR950321-100MG OR950321-250MG
APOSOR950321-100MGEA 3640 CZK
APOSOR950321-100MG APOSOR950321-250MG
1-(2-Bromobenzyl)-1H-imidazole ≥98%
1-(2-Bromobenzyl)-1H-imidazole
Vzorec: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD13396249
CAS číslo: 72459-45-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR