Tisk…

2-(2-Methylphenyl)propanenitrile 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(o-Tolyl)propanenitrile

OR950184-1G OR950184-250MG
APOSOR950184-1GEA 2240 CZK
APOSOR950184-1G APOSOR950184-250MG
2-(2-Methylphenyl)propanenitrile 95%
2-(2-Methylphenyl)propanenitrile
Vzorec: C₁₀H₁₁N
MW: 145,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036533
CAS číslo: 58422-60-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR