3-Chloropyridine-4-carboxylic acid ethyl ester 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 3-chloroisonicotinate
OR950143-5G
APOSOR950143-5GEA
2650
CZK
APOSOR950143-5G
3-Chloropyridine-4-carboxylic acid ethyl ester 95%
3-Chloropyridine-4-carboxylic acid ethyl ester
Vzorec:
C₈H₈ClNO₂ MW: 185,608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411706 CAS číslo: 211678-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...