Tisk…

N,N-Dimethyl-2-(p-tolyloxy)ethanamine 95%

Dodavatel: Apollo Scientific

OR950121-1G OR950121-250MG
APOSOR950121-1GEA 2320 CZK
APOSOR950121-1G APOSOR950121-250MG
N,N-Dimethyl-2-(p-tolyloxy)ethanamine 95%
N,N-Dimethyl-2-(p-tolyloxy)ethanamine
Vzorec: C₁₁H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD06304626
CAS číslo: 51344-14-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR