Tisk…

(2-Methyl-6-nitrophenyl)methanol 95%

Dodavatel: Apollo Scientific

OR950008-1G OR950008-250MG
APOSOR950008-1GEA 11410 CZK
APOSOR950008-1G APOSOR950008-250MG
(2-Methyl-6-nitrophenyl)methanol 95%
(2-Methyl-6-nitrophenyl)methanol
Vzorec: C₈H₉NO₃
MW: 167,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031847
CAS číslo: 54915-41-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR