Tisk…

4-Bromoquinoline-6-carbonitrile 97%

Dodavatel: Apollo Scientific
Varování

OR949972-100MG OR949972-250MG
APOSOR949972-100MGEA 2030 CZK
APOSOR949972-100MG APOSOR949972-250MG
4-Bromoquinoline-6-carbonitrile 97%
4-Bromoquinoline-6-carbonitrile
Vzorec: C₁₀H₅BrN₂
MW: 233,064 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09261095
CAS číslo: 642477-82-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR