5-Chloro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
Dodavatel: Apollo Scientific
OR949942-1G
OR949942-250MG
APOSOR949942-1GEA
16960
CZK
APOSOR949942-1G
APOSOR949942-250MG
5-Chloro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
5-Chloro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₇H₇ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD18802343 CAS číslo: 1146970-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...