Tisk…

4-Oxocyclohexyl acetate 95%

Dodavatel: Apollo Scientific

OR949851-1G OR949851-250MG
APOSOR949851-1GEA 14250 CZK
APOSOR949851-1G APOSOR949851-250MG
4-Oxocyclohexyl acetate 95%
4-Oxocyclohexyl acetate
Vzorec: C₈H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD07779439
CAS číslo: 41043-88-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR