Tisk…

7-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Dodavatel: Apollo Scientific

OR949523-1G OR949523-250MG
APOSOR949523-1GEA 8600 CZK
APOSOR949523-1G APOSOR949523-250MG
7-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
7-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Vzorec: C₁₀H₁₃NO·ClH
MW: 199,677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01444681
CAS číslo: 1745-05-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR