Tisk…
1-{2,6-Diazaspiro[3.3]heptan-2-yl}ethan-1-one hydrochloride
Dodavatel: Apollo Scientific
OR949457-1G OR949457-250MG OR949457-500MG OR949457-5G OR949457-10G
APOSOR949457-1GEA 20470 CZK
APOSOR949457-1G APOSOR949457-250MG APOSOR949457-500MG APOSOR949457-5G APOSOR949457-10G
1-{2,6-Diazaspiro[3.3]heptan-2-yl}ethan-1-one hydrochloride
1-{2,6-Diazaspiro[3.3]heptan-2-yl}ethan-1-one hydrochloride
Vzorec: C₇H₁₂N₂O·ClH
MW: 176,644 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24619367
CAS číslo: 1349875-74-6