3-(Dimethylamino)cyclobutanecarboxylic acid
Dodavatel: Apollo Scientific
OR949445-1G
OR949445-250MG
OR949445-500MG
APOSOR949445-1GEA
28800
CZK
APOSOR949445-1G
APOSOR949445-250MG
APOSOR949445-500MG
3-(Dimethylamino)cyclobutanecarboxylic acid
3-(Dimethylamino)cyclobutanecarboxylic acid
Vzorec:
C₇H₁₃NO₂ MW: 143,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24531429 CAS číslo: 1628252-12-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...