Tisk…
cis-Methyl 2,2-dimethyl-3-hydroxycyclobutanecarboxylate
Dodavatel: Apollo Scientific
Synonyms: (1R,3S)-rel-Methyl 3-hydroxy-2,2-dimethylcyclobutanecarboxylate
OR949364-1G OR949364-250MG
APOSOR949364-1GEA 20900 CZK
APOSOR949364-1G APOSOR949364-250MG
cis-Methyl 2,2-dimethyl-3-hydroxycyclobutanecarboxylate
cis-Methyl 2,2-dimethyl-3-hydroxycyclobutanecarboxylate
Vzorec: C₈H₁₄O₃
MW: 158.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106162
CAS číslo: 1392804-41-9