Tisk…

1,7-Dihydro-pyrano[3,4-b]pyrrol-4-one 95%

Dodavatel: Apollo Scientific

OR949155-1G OR949155-5G
APOSOR949155-1GEA 5000 CZK
APOSOR949155-1G APOSOR949155-5G
1,7-Dihydro-pyrano[3,4-b]pyrrol-4-one 95%
1,7-Dihydro-pyrano[3,4-b]pyrrol-4-one
Vzorec: C₇H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD22071242
CAS číslo: 1369248-13-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR