Tisk…

6-Methylpyridine-2-boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Borono-6-methylpyridine, (6-Methylpyridin-2-yl)boronic acid

OR9485-1G OR9485-250MG
APOSOR9485-1GEA 11740 CZK
APOSOR9485-1G APOSOR9485-250MG
6-Methylpyridine-2-boronic acid
6-Methylpyridine-2-boronic acid
Vzorec: C₆H₈BNO₂
MW: 136.944 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03093331
CAS číslo: 372963-50-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR