Tisk…

1-(6-Chloropyridazin-3-yl)ethan-1-one

Dodavatel: Apollo Scientific

OR948492-1G OR948492-5G
APOSOR948492-1GEA 8010 CZK
APOSOR948492-1G APOSOR948492-5G
1-(6-Chloropyridazin-3-yl)ethan-1-one
1-(6-Chloropyridazin-3-yl)ethan-1-one
Vzorec: C₆H₅ClN₂O
MW: 156,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698042
CAS číslo: 214701-31-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR