4-Hydroxynicotinaldehyde 98%
Dodavatel: Apollo Scientific
OR948489-1G
OR948489-250MG
APOSOR948489-1GEA
8100
CZK
APOSOR948489-1G
APOSOR948489-250MG
4-Hydroxynicotinaldehyde 98%
4-Hydroxynicotinaldehyde
Vzorec:
C₆H₅NO₂ MW: 123,109 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10697542 CAS číslo: 89380-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...