3-Iodo-1H-indazole-6-carbaldehyde ≥98%
Dodavatel: Apollo Scientific
OR948475-100MG
OR948475-1G
OR948475-250MG
APOSOR948475-100MGEA
16830
CZK
APOSOR948475-100MG
APOSOR948475-1G
APOSOR948475-250MG
3-Iodo-1H-indazole-6-carbaldehyde ≥98%
3-Iodo-1H-indazole-6-carbaldehyde
Vzorec:
C₈H₅IN₂O MW: 272.043 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10696810 CAS číslo: 944904-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...