Tisk…

1-Methyl-1H-indazole-3-carbonitrile 95%

Dodavatel: Apollo Scientific

OR948461-1G OR948461-250MG
APOSOR948461-1GEA 18420 CZK
APOSOR948461-1G APOSOR948461-250MG
1-Methyl-1H-indazole-3-carbonitrile 95%
1-Methyl-1H-indazole-3-carbonitrile
Vzorec: C₉H₇N₃
MW: 157,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971307
CAS číslo: 31748-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR