Tisk…

Boc-(S)-2-Cyclobutylglycine 97%

Dodavatel: Apollo Scientific

OR948354-1G OR948354-250MG OR948354-500MG
APOSOR948354-1GEA 18620 CZK
APOSOR948354-1G APOSOR948354-250MG APOSOR948354-500MG
Boc-(S)-2-Cyclobutylglycine 97%
Boc-(S)-2-Cyclobutylglycine
Vzorec: C₁₁H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD06659975
CAS číslo: 155905-77-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR