Tisk…

4-Bromo-6-methylpyridin-2-amine 95%

Dodavatel: Apollo Scientific

OR948239-1G OR948239-250MG
APOSOR948239-1GEA 4630 CZK
APOSOR948239-1G APOSOR948239-250MG
4-Bromo-6-methylpyridin-2-amine 95%
4-Bromo-6-methylpyridin-2-amine
Vzorec: C₆H₇BrN₂
MW: 187,037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062948
CAS číslo: 524718-27-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR