Tisk…

b-Amino-4-bromobenzeneethanol 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-2-(4-bromophenyl)ethanol

OR948229-1G
APOSOR948229-1GEA 2240 CZK
APOSOR948229-1G
b-Amino-4-bromobenzeneethanol 97%
b-Amino-4-bromobenzeneethanol
Vzorec: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12782005
CAS číslo: 201403-02-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR