(S)-b-Amino-3-bromo-benzeneethanol 97%
Dodavatel: Apollo Scientific
Synonyms:
(S)-2-Amino-2-(3-bromophenyl)ethanol
OR948198-1G
OR948198-250MG
APOSOR948198-1GEA
4400
CZK
APOSOR948198-1G
APOSOR948198-250MG
(S)-b-Amino-3-bromo-benzeneethanol 97%
(S)-b-Amino-3-bromo-benzeneethanol
Vzorec:
C₈H₁₀BrNO MW: 216,075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09253669 CAS číslo: 209963-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...