(R)-b-Amino-4-bromo-benzeneethanol 97%
Dodavatel: Apollo Scientific
Synonyms:
(R)-2-Amino-2-(4-bromophenyl)ethanol
OR948197-1G
OR948197-250MG
APOSOR948197-1GEA
3910
CZK
APOSOR948197-1G
APOSOR948197-250MG
(R)-b-Amino-4-bromo-benzeneethanol 97%
(R)-b-Amino-4-bromo-benzeneethanol
Vzorec:
C₈H₁₀BrNO MW: 216,075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09256373 CAS číslo: 354153-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...