2-Methoxy-2-methylpropan-1-ol 95%
OR948084-1G
OR948084-250MG
APOSOR948084-1GEA
14290
CZK
APOSOR948084-1G
APOSOR948084-250MG
2-Methoxy-2-methylpropan-1-ol 95%
2-Methoxy-2-methylpropan-1-ol
Vzorec:
C₅H₁₂O₂ MW: 104,148 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18253497 CAS číslo: 22665-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...