3H-Imidazo[4,5-c]pyridin-2-ol 95%
Dodavatel: Apollo Scientific
Synonyms:
1H-Imidazo[4,5-c]pyridin-2(3H)-one
OR947396-5G
APOSOR947396-5GEA
4370
CZK
APOSOR947396-5G
3H-Imidazo[4,5-c]pyridin-2-ol 95%
3H-Imidazo[4,5-c]pyridin-2-ol
Vzorec:
C₆H₅N₃O MW: 135,123 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18872694 CAS číslo: 7397-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...