Tisk…

5-Formyl-2-methylbenzoic acid 95%

Dodavatel: Apollo Scientific

OR947362-1G OR947362-250MG
APOSOR947362-1GEA 18420 CZK
APOSOR947362-1G APOSOR947362-250MG
5-Formyl-2-methylbenzoic acid 95%
5-Formyl-2-methylbenzoic acid
Vzorec: C₉H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD26673176
CAS číslo: 105650-34-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR