3-Chloro-4-formylbenzonitrile 97%
Dodavatel: Apollo Scientific
OR947339-1G
OR947339-5G
APOSOR947339-1GEA
2860
CZK
APOSOR947339-1G
APOSOR947339-5G
3-Chloro-4-formylbenzonitrile 97%
3-Chloro-4-formylbenzonitrile
Vzorec:
C₈H₄ClNO MW: 165,576 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11036376 CAS číslo: 58588-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...