Tisk…

1-(tert-Butoxy)-4-methylbenzene 95%

Dodavatel: Apollo Scientific

OR947321-1G OR947321-5G
APOSOR947321-1GEA 870 CZK
APOSOR947321-1G APOSOR947321-5G
1-(tert-Butoxy)-4-methylbenzene 95%
1-(tert-Butoxy)-4-methylbenzene
Vzorec: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD03425268
CAS číslo: 15359-98-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR