Tisk…

3-(Methylamino)propanal, N-BOC protected 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl methyl(3-oxopropyl)carbamate, tert-Butyl N-methyl-N-(3-oxopropyl)carbamate

OR947154-1G OR947154-250MG
APOSOR947154-1GEA 12900 CZK
APOSOR947154-1G APOSOR947154-250MG
3-(Methylamino)propanal, N-BOC protected 95%
3-(Methylamino)propanal, N-BOC protected
Vzorec: C₉H₁₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD09751791
CAS číslo: 273757-11-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR