tert-Butyl 3-hydroxy-2-methylpropylcarbamate 95%
Dodavatel: Apollo Scientific
OR947144-1G
OR947144-250MG
APOSOR947144-1GEA
21490
CZK
APOSOR947144-1G
APOSOR947144-250MG
tert-Butyl 3-hydroxy-2-methylpropylcarbamate 95%
tert-Butyl 3-hydroxy-2-methylpropylcarbamate
Vzorec:
C₉H₁₉NO₃ MW: 189,252 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14635738 CAS číslo: 480451-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...