3-(Methylamino)cyclohexanol 95%
Dodavatel: Apollo Scientific
OR947031-1G
OR947031-250MG
APOSOR947031-1GEA
19010
CZK
APOSOR947031-1G
APOSOR947031-250MG
3-(Methylamino)cyclohexanol 95%
3-(Methylamino)cyclohexanol
Vzorec:
C₇H₁₅NO MW: 129,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16620320 CAS číslo: 89854-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...