5-Methoxynaphthalen-1-ol 95%
Dodavatel: Apollo Scientific
OR947028-1G
OR947028-250MG
APOSOR947028-1GEA
21490
CZK
APOSOR947028-1G
APOSOR947028-250MG
5-Methoxynaphthalen-1-ol 95%
5-Methoxynaphthalen-1-ol
Vzorec:
C₁₁H₁₀O₂ MW: 174,196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00512042 CAS číslo: 3588-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...