Tisk…

2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanol 95%

Dodavatel: Apollo Scientific

OR946864-1G OR946864-250MG
APOSOR946864-1GEA 20900 CZK
APOSOR946864-1G APOSOR946864-250MG
2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanol 95%
2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanol
Vzorec: C₈H₉N₃O
MW: 163,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00957173
CAS číslo: 938-56-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR