Benzyl[2-(methylamino)ethyl]amine 95%
Dodavatel: Apollo Scientific
OR946836-1G
OR946836-250MG
APOSOR946836-1GEA
22520
CZK
APOSOR946836-1G
APOSOR946836-250MG
Benzyl[2-(methylamino)ethyl]amine 95%
Benzyl[2-(methylamino)ethyl]amine
Vzorec:
C₁₀H₁₆N₂ Storage Temperature: Ambient |
MDL Number:
MFCD09037852 CAS číslo: 56904-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...