Tisk…

N-[4-(2-Hydroxyethyl)phenyl]acetamide 95%

Dodavatel: Apollo Scientific

Synonyms: N-(4-(2-Hydroxyethyl)phenyl)acetamide

OR946802-1G OR946802-5G
APOSOR946802-1GEA 2610 CZK
APOSOR946802-1G APOSOR946802-5G
N-[4-(2-Hydroxyethyl)phenyl]acetamide 95%
N-[4-(2-Hydroxyethyl)phenyl]acetamide
Vzorec: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01536093
CAS číslo: 83345-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR