Tisk…

2-Methyl-homopiperazine, N1-BOC protected 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Methyl-1,4-diazepane, N1-BOC protected

OR946706-1G OR946706-250MG
APOSOR946706-1GEA 31020 CZK
APOSOR946706-1G APOSOR946706-250MG
2-Methyl-homopiperazine, N1-BOC protected 98%
2-Methyl-homopiperazine, N1-BOC protected
Vzorec: C₁₁H₂₂N₂O₂
MW: 214,305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18432596
CAS číslo: 1260422-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR